Methyl oleate, Supelco®
Leverantör: Merck
Synonyms:
cis-Oleic Acid Methyl Ester, Oleic acid methyl ester
M-Clarity™ quality level = MQ100
75160-1ML
75160-5ML
75160-1MLEA
907
SEK
75160-1ML
75160-5ML
Methyl oleate, Supelco®
Methyl oleate
Formel:
C₁₉H₃₆O₂ MW: 296,49 g/mol Kokpunkt: ∼218,5 °C (1013 hPa) Smältpunkt: –20 °C Densitet: 0,88 g/cm³ (20 °C) Storage Temperature: Omgivande |
MDL Number:
MFCD00009578 CAS nummer: 112-62-9 EINECS: 203-992-5 Merck Index: 12,06965 |