3-Klorpropen ≥98% stabilised
Leverantör: Thermo Scientific
Fara
Synonyms: 3-Klor-1-propen
Stabilization: Stabilised with propylene oxide
A14330.0F A14330.AP
A14330.0FEA 2525 SEK
A14330.0F A14330.AP
3-Klorpropen ≥98% stabilised
3-Klorpropen
Stabilized with propylene oxide
Purity: 98% CAS No.: 107-05-1 Molecular Formula: C3H5Cl Formula Weight: 76.53 Storage Temperature: 2°C to 8°C Flash Point: -28°(-18°F) Refractive Index: 1.4155 Odour: Sweet Melting Point: -135° Boiling Point: 44-46° Beilstein Reference: 635704 Merck Reference: 14,289 UN No.: 1100 MDL No.: MFCD00000984
Formel: C₃H₅Cl
MW: 76,53 g/mol
Kokpunkt: 44…46 °C
Smältpunkt: –135 °C
Densitet: 0,939
Flampunkt: –28 °C (–18 °F)
Storage Temperature: Kylskåp
MDL Number: MFCD00000984
CAS nummer: 107-05-1
EINECS: 203-457-6
UN: 1100
ADR: 3,I
Merck Index: 14,289