HEPES sodium salt
Leverantör: Molekula
Synonyms: Sodium 4-(2-hydroxyethyl)piperazin-1-ylethanesulphonate, Sodium 2-(4-(2-hydroxyethyl)piperazin-1-yl)ethanesulphonate, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulphonic acid) sodium salt, 4-(2-Hydroxyethyl)piperazine-1-ethanesulphonic acid sodium salt, HEPES sodium, HEPES Na
80267503-500G 80267503-100G
MOLE80267503-500GEA 0 SEK
MOLE80267503-500G MOLE80267503-100G
HEPES sodium salt
HEPES sodium salt
Formel: C₈H₁₇N₂NaO₄S
MW: 260,29 g/mol
Smältpunkt: ? 105 °C
Storage Temperature: Omgivande
MDL Number: MFCD00036463
CAS nummer: 75277-39-3
EINECS: 278-169-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=116e9b4f3c17d013ff2b7c9848c219943a725b3c&vid=6a897b63469c2540240df1cc349c2773d57bcdaa&prid=web&clid=SAPDAM