2-[4 (2-Hydroxietyl)-1-piperazinyl]-ethan sulfonsyra, pulver cell culture reagent, not sterile
Leverantör: BIOWEST
Synonyms: HEPES free acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulphonic acid), 4-(2-Hydroxyethyl)piperazin-1-ylethanesulphonic acid, N-2-Hydroxyethylpiperazine-N′-2-ethanesulphonic acid, 2-(4-(2-Hydroxyethyl)piperazin-1-yl)ethanesulphonic acid
P5455-250GR P5455-500GR P5455-1KG
P5455-250GREA 1250 SEK
P5455-250GR P5455-500GR P5455-1KG
2-[4 (2-Hydroxietyl)-1-piperazinyl]-ethan sulfonsyra, pulver cell culture reagent, not sterile
HEPES
Formel: C₈H₁₈N₂O₄S
MW: 238.31 g/mol
Smältpunkt: 210…215 °C
Densitet: 1.448 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00006158
CAS nummer: 7365-45-9
EINECS: 230-907-9
Merck Index: 13,04672
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4cdc09c866676149941b93b2cac8d453175059a2&vid=0b09e5b84ea34a806443c3071fad63e15e4e3433&prid=web&clid=SAPDAM