(Pentafluorophenyl)acetic acid ≥98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 2,3,4,5,6-Pentafluorophenylacetic acid
B22430.03 B22430.06
B22430.03EA 1050 SEK
B22430.03 B22430.06
(Pentafluorophenyl)acetic acid ≥98%
(Pentafluorophenyl)acetic acid
Purity: 98+% CAS No.: 653-21-4 Molecular Formula: C8H3F5O2 Formula Weight: 226.10 Melting Point: 107-110° Beilstein Reference: 2056612
Formel: C₈H₃F₅O₂
MW: 226.1 g/mol
Kokpunkt: 247
Smältpunkt: 107…110 °C
Storage Temperature: Omgivande
MDL Number: MFCD00004316
CAS nummer: 653-21-4
EINECS: 211-497-0