(4-Methoxyphenyl)acetic acid 99%
Leverantör: Thermo Scientific
Varning
Synonyms: 4-Methoxyphenylacetic acid
126020050 126021000 126022500 126020010
ACRO126020050EA 440 SEK
ACRO126020050 ACRO126021000 ACRO126022500 ACRO126020010
(4-Methoxyphenyl)acetic acid 99%
(4-Methoxyphenyl)acetic acid
Formel: (C₆H₄(OCH₃))CH₂COOH
MW: 166,18 g/mol
Kokpunkt: 140 °C (3 mmHg)
Smältpunkt: 84…88 °C
Flampunkt: 193 °C
Storage Temperature: Omgivande
MDL Number: MFCD00004345
CAS nummer: 104-01-8
EINECS: 203-166-4
Order Now
Specifikation testresultat
Appearance White to light yellow Crystalline powder and/or chunks
Infrared spectrum Conforms
Melting point 84 to 88 °C
Titration with NaOH ≥98.5 %