(4-Nitrophenyl)acetic acid 99%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 4-Nitrophenylacetic acid
128760500 128761000
ACRO128760500EA 735 SEK
ACRO128760500 ACRO128761000
(4-Nitrophenyl)acetic acid 99%
(4-Nitrophenyl)acetic acid
Formel: (C₆H₄(NO₂))CH₂COOH
MW: 181.15 g/mol
Smältpunkt: 152…153 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007383
CAS nummer: 104-03-0
EINECS: 203-168-5
Merck Index: 12,06719
Order Now
Specifikation testresultat
Appearance Yellow to beige Crystalline powder
Infrared spectrum Conforms
Melting point 153 to 156 °C
HPLC ≥98.5 %
Titration with NaOH ≥98.5 %