4-Nitrophenyl chloroformate 97%
Leverantör: Thermo Scientific
Fara
170800050 170800250 170801000 170805000
ACRO170800050EA 670 SEK
ACRO170800050 ACRO170800250 ACRO170801000 ACRO170805000
4-Nitrophenyl chloroformate 97%
4-Nitrophenyl chloroformate
Formel: C₇H₄ClNO₄
MW: 201,57 g/mol
Kokpunkt: 159…162 °C (19 mmHg)
Smältpunkt: 73…81 °C
Flampunkt: >110 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00007321
CAS nummer: 7693-46-1
EINECS: 231-706-9
UN: 2923
ADR: 8,II
Order Now
Specifikation testresultat
Appearance White to yellow to light beige Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 77 to 82 °C
Titration Argentometric ≥96.0 % (ex Cl)