(3,4-Dichlorophenyl)acetic acid ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: 3,4-Dichlorophenylacetic acid
A15653.06 A15653.14 A15653.22
A15653.06EA 645 SEK
A15653.06 A15653.14 A15653.22
(3,4-Dichlorophenyl)acetic acid ≥98%
(3,4-Dichlorophenyl)acetic acid
Purity: 98% CAS No.: 5807-30-7 Molecular Formula: C8H6Cl2O2 Formula Weight: 205.04 Odour: Odourless Melting Point: 87-92° Beilstein Reference: 1953781 MDL No.: MFCD00004333
Formel: (C₆H₃Cl₂)CH₂COOH
MW: 205,04 g/mol
Kokpunkt: 314 °C (1013 hPa)
Smältpunkt: 87…92 °C
Storage Temperature: Omgivande
MDL Number: MFCD00004333
CAS nummer: 5807-30-7
EINECS: 227-368-7