Indol-3-acetic syra, Millipore®
Leverantör: Merck
Synonyms: IAA, 1H-Indole-3-acetic acid, Heteroauxin, 3-Indoleacetic acid
M-Clarity™ quality level = MQ100
1.00353.0010 1.00353.0100
1.00353.0010EA 755 SEK
1.00353.0010 1.00353.0100
Indol-3-acetic syra, Millipore®
Indol-3-acetic syra
Formel: C₁₀H₉NO₂
MW: 175,19 g/mol
Kokpunkt: 120…125 °C (0,05 mmHg)
Smältpunkt: 167…170 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00005636
CAS nummer: 87-51-4
EINECS: 201-748-2
Merck Index: 13,04986