Indol-3-acetic syra ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: IAA, 1H-Indole-3-acetic acid, Heteroauxin, 3-Indoleacetic acid
A10556.06 A10556.14 A10556.22 A10556.36
A10556.06EA 640 SEK
A10556.06 A10556.14 A10556.22 A10556.36
Indol-3-acetic syra ≥98%
Indol-3-acetic syra
Purity: 98+% CAS No.: 87-51-4 Molecular Formula: C10H9NO2 Formula Weight: 175.19 Storage Temperature: 2°C to 8°C Flash Point: 171°(339°F) Melting Point: ca 167° dec. Beilstein Reference: 143358 Merck Reference: 14,4964 MDL No.: MFCD00005636
Formel: C₁₀H₉NO₂
MW: 175,19 g/mol
Kokpunkt: 120…125 °C (0,05 mmHg)
Smältpunkt: ∼ 167 °C (dec.)
Flampunkt: 171 °C (339 °F)
Storage Temperature: Kylskåp
MDL Number: MFCD00005636
CAS nummer: 87-51-4
EINECS: 201-748-2
Merck Index: 14,4964