2-Naphthylacetic acid 99%
Leverantör: Thermo Fisher Scientific
Varning
128220050 128220250 128221000
ACRO128220050EA 795 SEK
ACRO128220050 ACRO128220250 ACRO128221000
2-Naphthylacetic acid 99%
2-Naphthylacetic acid
Formel: C₁₂H₁₀O₂
Kokpunkt: 352 °C (1013 hPa)
Smältpunkt: 141…143 °C
Densitet: 1,29 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00004126
CAS nummer: 581-96-4
EINECS: 209-475-0
Order Now
Specifikation testresultat
Appearance White to off-white Crystalline powder or flakes
Infrared spectrum Conforms
Melting point 141 to 143 °C
GC ≥98.5 %
Titration with NaOH ≥98.5 %