2-Metylbutan ≥99%, Reagent Grade
Leverantör: Honeywell Chemicals
Fara
Synonyms: Isopentan
M32631-1L M32631-2.5L M32631-6X1L M32631-500ML
M32631-1LEA 985 SEK
M32631-1L M32631-2.5L M32631-4X1L M32631-500ML
2-Metylbutan ≥99%, Reagent Grade
2-Metylbutan
  • ≥95% for general laboratory use.
Beilstein Number: 1730723
RTECS Number: EK4430000
UNSPSC Code: 12191502
Formel: CH₃CH₂CH(CH₃)₂
MW: 72,15 g/mol
Kokpunkt: 7,6 %(V)
Smältpunkt: -160 °C
Densitet: 0,62 g/cm³ (20 °C)
Flampunkt: <-35 °C
Storage Temperature: Omgivande
MDL Number: MFCD00009338
CAS nummer: 78-78-4
EINECS: 201-142-8
UN: 1265
ADR: 3,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=498904a9f8b2618e19d2343aa74e247ec7121f00&vid=278ab1939f4cb13ad32cc36a860f8e72113575ba&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Vapor Pressure 11.17 psi ( 20 °C)
Identity (IR) complying
Assay (GC) Min. 99.0 %
Autoignition Temperature 788 °F
Explosion Limit 8.3 %
Refractive Index n20/D 1.354 (lit.)
Vapor Density 2.6 (vs air)