2-Metyl-2-propanol, EMPLURA®, Supelco®
Leverantör: Merck
|
Fara
|
M-Clarity™ quality level = MQ200
8.22264.1000
8.22264.9025
8.22264.1000EA
813
SEK
8.22264.1000
8.22264.9025
2-Metyl-2-propanol, EMPLURA®, Supelco®
2-Metyl-2-propanol
Formel:
(CH₃)₃COH Kokpunkt: 82,2 °C (1013 hPa) Densitet: 0,7887 g/cm³ (20 °C) Flampunkt: 11 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00004464 CAS nummer: 75-65-0 EINECS: 200-889-7 UN: 1120 ADR: 3,II Merck Index: 12,01577 |