2,2-Dimethyl-1-propanol ≥99%
Leverantör: Thermo Scientific
Fara
Synonyms: Neopentyl alcohol
A14390.14 A14390.22 A14390.36
A14390.14EA 835 SEK
A14390.14 A14390.22 A14390.36
2,2-Dimethyl-1-propanol ≥99%
2,2-Dimethyl-1-propanol
Purity: 99% CAS No.: 75-84-3 Molecular Formula: (CH3)3CCH2OH Formula Weight: 88.15 Flash Point: 36°(96°F) Melting Point: 52-56° Boiling Point: 111-114° Beilstein Reference: 1730984 Merck Reference: 14,6457 UN No.: 1325 MDL No.: MFCD00004682
Formel: C₅H₁₂O
MW: 88,15 g/mol
Kokpunkt: 111…114 °C
Smältpunkt: 52…56 °C
Densitet: 0,818
Flampunkt: 36 °C (96 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00004682
CAS nummer: 75-84-3
EINECS: 200-907-3
UN: 1325
ADR: 4.1,II
Merck Index: 14,6457