Metyl-tert-butyleter, extra dry 99+%, AcroSeal®
Leverantör: Thermo Fisher Scientific
Fara
375211000 375210010
375211000.EA 825 SEK
375211000. 375210010.
Metyl-tert-butyleter, extra dry 99+%, AcroSeal®
Metyl-tert-butyleter
Formel: (CH₃)₃COCH₃
Kokpunkt: 54…56 °C (760 mmHg)
Smältpunkt: –110 °C
Densitet: 0,7404 g/cm³
Flampunkt: –28 °C
Storage Temperature: Omgivande
MDL Number: MFCD00008812
CAS nummer: 1634-04-4
EINECS: 216-653-1
UN: 2398
ADR: 3,II
Merck Index: 13,06059
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1712bc714ba0a06e0de37f7cf4e5117256f84425&vid=e1b314f829136075329c50f9e5a593c19ae28b4f&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 %
GC ≥99.0 %
Residue after evaporation ≤0.002 %
Peroxides ≤0.005 % (as H2O2)