Isopentansyra ≥98%
Leverantör: Thermo Scientific
Fara
Synonyms: 3-Methylbutyric acid, Isopropyl acetic acid, 3-Methylbutanoic acid
A18642.AK A18642.AU
A18642.AKEA 705 SEK
A18642.AK A18642.AU
Isopentansyra ≥98%
Isopentansyra
Purity: 98% CAS No.: 503-74-2 Molecular Formula: (CH3)2CHCH2CO2H Formula Weight: 102.13 Flash Point: 70°(158°F) Refractive Index: 1.4030 Odour: Rancid pH: 3 Melting Point: -33° Boiling Point: 175-177° Beilstein Reference: 1098522 Merck Reference: 14,5231 UN No.: 3265 MDL No.: MFCD00002726
Formel: C₅H₁₀O₂
MW: 102,13 g/mol
Kokpunkt: 175…177 °C
Smältpunkt: –33 °C
Densitet: 0,937
Flampunkt: 70 °C (158 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00002726
CAS nummer: 503-74-2
EINECS: 207-975-3
UN: 3265
ADR: 8,II
Merck Index: 14,5231