3,3-Dimethylbutyric acid 98%
Leverantör: Thermo Scientific
Fara
Synonyms: 3,3-Dimethylbutanoic acid, tert-Butylacetic acid
107770100 107770500 107772500
ACRO107770100EA 500 SEK
ACRO107770100 ACRO107770500 ACRO107772500
3,3-Dimethylbutyric acid 98%
3,3-Dimethylbutyric acid
Formel: C₆H₁₂O₂
MW: 116,16 g/mol
Kokpunkt: 185…190 °C (760 mmHg)
Smältpunkt: 8…6 °C
Densitet: 0,91 g/cm³
Flampunkt: 88 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002715
CAS nummer: 1070-83-3
EINECS: 213-982-2
UN: 3265
ADR: 8,III
Merck Index: 13,01537
Order Now
Specifikation testresultat
Appearance Yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4090 to 1.4110 (20°C, 589 nm)
Titration with NaOH ≥97.5 %