Isosmörsyra ≥99%
Leverantör: Thermo Scientific
Varning
Synonyms: 2-Metylpropionsyra
L04038.AE L04038.AP L04038.0F
L04038.AEEA 550 SEK
L04038.AE L04038.AP L04038.0F
Isosmörsyra ≥99%
Isosmörsyra
Purity: 99% CAS No.: 79-31-2 Molecular Formula: (CH3)2CHCO2H Formula Weight: 88.11 Flash Point: 59°(138°F) Refractive Index: 1.3928 Odour: Pungent Melting Point: -47° Boiling Point: 153-154° Beilstein Reference: 635770 Merck Reference: 14,5155 UN No.: 2529 MDL No.: MFCD00002658
Formel: C₄H₈O₂
MW: 88,11 g/mol
Kokpunkt: 153…154 °C
Smältpunkt: –47 °C
Densitet: 0,948
Flampunkt: 59 °C (138 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00002658
CAS nummer: 79-31-2
EINECS: 201-195-7
UN: 2529
ADR: 3,III
Merck Index: 14,5155