2-Etylhexansyra 99%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 2-Ethylhexanoic acid
118520025 118525000 118520011 118520100
ACRO118520025EA 1950 SEK
ACRO118520025 ACRO118525000 ACRO118520011 ACRO118520100
2-Etylhexansyra 99%
2-Etylhexansyra
Formel: C₈H₁₆O₂
MW: 144.21 g/mol
Kokpunkt: 228 °C (760 mmHg)
Smältpunkt: –59 °C
Densitet: 0.906 g/cm³
Flampunkt: 114 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002675
CAS nummer: 149-57-5
EINECS: 205-743-6
UN: 3265
ADR: 8 III
Order Now
Specifikation testresultat
Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4240 to 1.4260 (20°C, 589 nm)
Water ≤0.1 % (K.F.)
GC ≥98.5 %
Color scale ≤15 APHA
Iron (Fe) ≤5 ppm
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 %
SPECIFIKATIONER
Environmentally Preferable