2-Ethylbutyric acid ≥98%
Leverantör: Thermo Scientific
Fara
Synonyms: 2-Ethylbutanoic acid, Diethylacetic acid
A16989.AK A16989.AU
A16989.AKEA 1325 SEK
A16989.AK A16989.AU
2-Ethylbutyric acid ≥98%
2-Ethylbutyric acid
Purity: 98% CAS No.: 88-09-5 Molecular Formula: (CH3CH2)2CHCO2H Formula Weight: 116.16 Flash Point: 87°(188°F) Refractive Index: 1.4130 Melting Point: -15° Boiling Point: 192-194° Beilstein Reference: 1098634 Merck Reference: 14,3110 UN No.: 2922 MDL No.: MFCD00002670
Formel: C₆H₁₂O₂
MW: 116,16 g/mol
Kokpunkt: 192…194 °C
Smältpunkt: –15 °C
Densitet: 0,924
Flampunkt: 87 °C (188 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00002670
CAS nummer: 88-09-5
EINECS: 201-796-4
UN: 2810
ADR: 6.1,III
Merck Index: 14,3110