Pivalic acid zS, Sigma-Aldrich®
Leverantör: Merck
Fara
Synonyms: 2,2-Dimethylpropionic acid, Trimethylacetic acid
MQ200
8.01336.0100 8.01336.1000 8.01336.9050
8.01336.0100EA 577 SEK
8.01336.0100 8.01336.1000 8.01336.9050
Pivalic acid zS, Sigma-Aldrich®
Pivalic acid
Formel: C₅H₁₀O₂
MW: 102.13 g/mol
Kokpunkt: 164
Smältpunkt: 32…35 °C
Densitet: 0.91 g/cm³ (20 °C)
Flampunkt: 640
Storage Temperature: Omgivande
MDL Number: MFCD00004194
CAS nummer: 75-98-9
EINECS: 200-922-5
UN: 1759
ADR: 8 III
Merck Index: 12,07667
Order Now
Specifikation testresultat
Solubility 25 g/l (20 °C)
Melting Point 32 - 35 °C
Molar Mass 102.13 g/mol
Boiling Point 164 °C
Vapor Pressure 0.85 hPa (20 °C)
Flash Point 64 °C
Refractive Index 1.3931 (37 °C, 589 nm)
Explosion Limit 1.6 %(V)
Density 0.91 g/cm3 (20 °C)