Pivalic acid 99%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 2,2-Dimethylpropionic acid, Trimethylacetic acid
140020050 140021000 140025000 140020025
ACRO140020050EA 545 SEK
ACRO140020050 ACRO140021000 ACRO140025000 ACRO140020025
Pivalic acid 99%
Pivalic acid
Formel: C₅H₁₀O₂
MW: 102.13 g/mol
Kokpunkt: 163…164 °C (760 mmHg)
Smältpunkt: 33…35 °C
Flampunkt: 63 °C
Storage Temperature: Omgivande
MDL Number: MFCD00004194
CAS nummer: 75-98-9
EINECS: 200-922-5
Merck Index: 12,07667
Order Now
Specifikation testresultat
Appearance White to off-white Low melting crystalline mass
Infrared spectrum Conforms
GC ≥96.0 %