2,2-Dimethylbutyric acid ≥97%
Leverantör: Thermo Scientific
Fara
Synonyms: 2,2-Dimethylbutanoic acid
A17072.09 A17072.18 A17072.30
A17072.09EA 665 SEK
A17072.09 A17072.18 A17072.30
2,2-Dimethylbutyric acid ≥97%
2,2-Dimethylbutyric acid
Purity: 97% CAS No.: 595-37-9 Molecular Formula: C6H12O2 Formula Weight: 116.16 Flash Point: 90°(194°F) Refractive Index: 1.4154 Odour: Pungent Melting Point: -14° Boiling Point: 186° Beilstein Reference: 1720666 UN No.: 3265 MDL No.: MFCD00004200
Formel: C₆H₁₂O₂
MW: 116,16 g/mol
Kokpunkt: 186 °C
Smältpunkt: –14 °C
Densitet: 0,926
Flampunkt: 90 °C (194 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00004200
CAS nummer: 595-37-9
EINECS: 209-865-0
UN: 3265
ADR: 8,III