Ethyl pivalate ≥99%
Leverantör: Thermo Scientific
Fara
Synonyms: Trimethylacetic acid ethyl ester, Ethyltrimethylacetate, Pivalic acid ethyl ester, 2,2-Dimethylpropionic acid ethyl ester, Ethyl trimethylacetate
A13100.14 A13100.22
A13100.14EA 690 SEK
A13100.14 A13100.22
Ethyl pivalate ≥99%
Ethyl pivalate
Purity: 99% CAS No.: 3938-95-2 Molecular Formula: (CH3)3CCO2CH2CH3 Formula Weight: 130.19 Flash Point: 16°(60°F) Refractive Index: 1.3930 Odour: Stench Melting Point: -90° Boiling Point: 116-118° Beilstein Reference: 1747617 Merck Reference: 14,7511 UN No.: 3272 MDL No.: MFCD00009129
Formel: C₇H₁₄O₂
MW: 130,19 g/mol
Kokpunkt: 116…118 °C
Smältpunkt: –90 °C
Densitet: 0,891
Flampunkt: 16 °C (60 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00009129
CAS nummer: 3938-95-2
EINECS: 223-520-1
UN: 3272
ADR: 3,II
Merck Index: 14,7511