L(-)-Glutathione (oxidised form)
Leverantör: PanReac AppliChem
Synonyms: L(-)-Glutathione, oxidized, Bi(glutathion-S-yl), GSSG, Glutathiol, Oxidized Glutathione (GSSG), Glutathione oxidized, (2S,2'S)-5,5'-(((2R,2'R)-Disulphanediylbis(1-((carboxymethyl)amino)-1-oxopropane-3,2-diyl))bis(azanediyl))bis(2-amino-5-oxopentanoic acid)
A2243.0005 A2243.0025
A2243.0005EA 2850 SEK
A2243.0005 A2243.0025
L(-)-Glutathione (oxidised form)
L(-)-Glutathione (oxidised form)
Formel: C₂₀H₃₂N₆O₁₂S₂
MW: 612.64 g/mol
Smältpunkt: 185
Storage Temperature: Kylskåp
MDL Number: MFCD00063106
CAS nummer: 27025-41-8
EINECS: 248-170-7