L (-)-Glutation (reducerad form), Millipore®
Leverantör: Merck Millipore Calbiochem
Synonyms: L-Glutathione Reduced, GSH, (S)-2-Amino-5-(((R)-1-((carboxymethyl)amino)-3-mercapto-1-oxopropan-2-yl)amino)-5-oxopentanoic acid, Glutathione Reduced
3541-1 3541-100 3541-25 3541-5
3541-1EA 655 SEK
3541-1 3541-100 3541-25 3541-5
L (-)-Glutation (reducerad form), Millipore®
L (-)-Glutation (reducerad form)
Formel: C₁₀H₁₇N₃O₆S
Smältpunkt: 185…195 °C
Densitet: 1,475 g/cm³ (20 °C)
Storage Temperature: Kylskåp
MDL Number: MFCD00065939
CAS nummer: 70-18-8
EINECS: 200-725-4
Merck Index: 13,04488