3-Hydroxy-3-methylbutanone ≥90%
Leverantör: Thermo Scientific
Varning
Synonyms: 3-Hydroxy-3-methyl-2-butanone, 3-Hydroxy-3-methylbutan-2-one
43724.22 43724.14 43724.06
43724.22EA 9700 SEK
43724.22 43724.14 43724.06
3-Hydroxy-3-methylbutanone ≥90%
3-Hydroxy-3-methylbutanone
Purity: 90+% CAS No.: 115-22-0 Molecular Formula: C5H10O2 Formula Weight: 102.13 Flash Point: 42°(107.6°F) Physical Form: Liquid Density: 0.96 Refractive Index: 1.4150 Boiling Point: 140-141° UN No.: 1224 MDL No.: MFCD00004460
Formel: C₅H₁₀O₂
MW: 102,13 g/mol
Kokpunkt: 140…141 °C
Densitet: 0,96
Flampunkt: 42 °C (107,6 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00004460
CAS nummer: 115-22-0
EINECS: 204-073-1
UN: 1224
ADR: 3,III