1-Bromo-3,3-dimethylbutanone 93%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 1-Bromopinacolone, 1-Bromo-3,3-dimethyl-2-butanone
306410050 306410250
ACRO306410050EA 970 SEK
ACRO306410050 ACRO306410250
1-Bromo-3,3-dimethylbutanone 93%
1-Bromo-3,3-dimethylbutanone
Formel: C₆H₁₁BrO
Kokpunkt: 189…192 °C (760 mmHg)
Smältpunkt: –10 °C
Densitet: 1,33 g/cm³
Flampunkt: 75 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00000206
CAS nummer: 5469-26-1
EINECS: 226-794-0
ADR: 6.1,II
Order Now
Specifikation testresultat
Appearance Clear light yellow to yellow-brown Liquid
Infrared spectrum Conforms
Refractive index 1.4656 to 1.4676 (20°C, 589 nm)
GC ≥91 %