Natriumhexan-1-sulfonatmonohydrat ≥98.0% för HPLC, för jonparkromatografi, J.T.Baker®
Leverantör: Avantor
Synonyms: Sodium hexane-1-sulphonate, Sodium hexanesulphonate, Sodium 1-hexanesulphonate
Management of Change (MOC) category = N
2175-05EA 2300 SEK
2175-05 2175-01
Natriumhexan-1-sulfonatmonohydrat ≥98.0% för HPLC, för jonparkromatografi, J.T.Baker®
Natriumhexan-1-sulfonatmonohydrat
Formel: C₆H₁₃O₃SNa
MW: 188,22 g/mol
Smältpunkt: 300 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007542
CAS nummer: 2832-45-3
EINECS: 220-601-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aebc3cb7a7cefac9ad5a7678d82a90d244b508f9&vid=f98fce9a53ea29283b11fb44162f26067930bbbe&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
For Laboratory,Research,or Manufacturing Use = 98.0 %
Assay (as C₆H₁₃SO₃Na) (by acidimetry) ≥98.0 %
UV Absorbance of 0.25 M Solution - 250 nm ≤0.050
UV Absorbance of 0.25 M Solution - 240 nm ≤0.050
UV Absorbance of 0.25 M Solution - 230 nm ≤0.050
UV Absorbance of 0.25 M Solution - 220 nm ≤0.060
UV Absorbance of 0.25 M Solution - 210 nm ≤0.080
UV Absorbance of 0.25 M Solution - 200 nm ≤0.200
Appearance (clear, colorless, mobile liquid)