Natriumhexan-1-sulfonatmonohydrat ≥99%
Leverantör: Thermo Scientific
Synonyms: Sodium hexane-1-sulphonate, Sodium hexanesulphonate, Sodium 1-hexanesulphonate
A13278.14 A13278.06 A13278.22
A13278.14EA 4175 SEK
A13278.14 A13278.06 A13278.22
Natriumhexan-1-sulfonatmonohydrat ≥99%
Natriumhexan-1-sulfonatmonohydrat
Purity: 99% CAS No.: 2832-45-3 Molecular Formula: CH3(CH2)5SO3Na Formula Weight: 188.22 Physical Form: Powder Melting Point: 300° Beilstein Reference: 3727014 MDL No.: MFCD00007542
Formel: C₆H₁₃O₃SNa
MW: 188,22 g/mol
Smältpunkt: >300 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007542
CAS nummer: 2832-45-3
EINECS: 220-601-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aebc3cb7a7cefac9ad5a7678d82a90d244b508f9&vid=f98fce9a53ea29283b11fb44162f26067930bbbe&prid=web&clid=SAPDAM