Natriumheptan-1-sulfonat, (max. 2% H₂O) 99%
Leverantör: Thermo Scientific
Synonyms: Sodium heptane-1-sulphonate, Sodium 1-heptanesulphonate
A10917.14 A10917.06 A10917.22
A10917.14EA 3725 SEK
A10917.14 A10917.06 A10917.22
Natriumheptan-1-sulfonat, (max. 2% H₂O) 99%
Natriumheptan-1-sulfonat
Purity: 99% CAS No.: 22767-50-6 Molecular Formula: CH3(CH2)6SO3Na Formula Weight: 202.24 Odour: Odourless Melting Point: 300° Beilstein Reference: 3731762 MDL No.: MFCD00007543
Formel: H₃C(CH₂)₅CH₂SO₃Na
MW: 202,25 g/mol
Smältpunkt: >300 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007543
CAS nummer: 22767-50-6
EINECS: 245-210-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=95c09a5691140bab7446d88919f01593e8f0b9b2&vid=ee549604b02a9d5e86a59503ed2dcf48a4c7cfd1&prid=web&clid=SAPDAM