1-Dodekansulfonsyra natriumsalt, Sigma-Aldrich®
Leverantör: Merck
Synonyms: Sodium 1-dodecanesulphonate, Dodecane-1-sulphonic acid sodium salt, Sodium 1-dodecansulphonate, 1-Dodecanesulphonic acid, sodium salt, Sodium dodecane-1-sulphonate
M-Clarity™ quality level = MQ200
1.12146.0005 1.12146.8500
1.12146.0005EA 1360 SEK
1.12146.0005 1.12146.8500
1-Dodekansulfonsyra natriumsalt, Sigma-Aldrich®
1-Dodekansulfonsyra natriumsalt
Formel: C₁₂H₂₅NaO₃S
MW: 272.38 g/mol
Smältpunkt: 300
Densitet: 1.218 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00007527
CAS nummer: 2386-53-0
EINECS: 219-200-6
Order Now
Specifikation testresultat
Solubility (20 °C) soluble
Molar Mass 272.37 g/mol
Bulk Density 330 kg/m3
pH 5.5 - 7.5 (100 g/l, H2O, 20 °C)