1-Butanesulphonic acid sodium salt, Supelco®
Leverantör: Merck
Synonyms: Sodium 1-butanesulphonate, Sodium butane-1-sulphonate, Sodium 1-butansulphonate, Butane-1-sulphonic acid sodium salt
M-Clarity™ quality level = MQ100
19022-10G-F 19022-50G-F
19022-10G-FEA 1760 SEK
19022-10G-F 19022-50G-F
1-Butanesulphonic acid sodium salt, Supelco®
1-Butanesulphonic acid sodium salt
Formel: C₄H₉NaO₃S
MW: 160.17 g/mol
Smältpunkt: > 300 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007540
CAS nummer: 2386-54-1
EINECS: 219-201-1