1-Butanesulphonic acid sodium salt ≥99%
Leverantör: Thermo Scientific
Synonyms: Sodium 1-butanesulphonate, Sodium butane-1-sulphonate, Sodium 1-butansulphonate, Butane-1-sulphonic acid sodium salt
A12654.06 A12654.14 A12654.22
A12654.06EA 1125 SEK
A12654.06 A12654.14 A12654.22
1-Butanesulphonic acid sodium salt ≥99%
1-Butanesulphonic acid sodium salt
Purity: 99% CAS No.: 2386-54-1 Molecular Formula: CH3(CH2)3SO3Na Formula Weight: 160.17 Melting Point: 300° Beilstein Reference: 3716662
Formel: C₄H₉NaO₃S
MW: 160,17 g/mol
Smältpunkt: >300 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007540
CAS nummer: 2386-54-1
EINECS: 219-201-1