Ethanesulphonyl chloride ≥98%
Leverantör: Thermo Scientific
Fara
B23723.14 B23723.22 B23723.36
B23723.14EA 565 SEK
B23723.14 B23723.22 B23723.36
Ethanesulphonyl chloride ≥98%
Ethanesulphonyl chloride
Purity: 98+% CAS No.: 594-44-5 Molecular Formula: C2H5ClO2S Formula Weight: 128.57 Flash Point: 83°(181°F) Refractive Index: 1.4540 Melting Point: -70° Boiling Point: 176-178° Beilstein Reference: 773865 UN No.: 2927 MDL No.: MFCD00007460
Formel: C₂H₅ClO₂S
MW: 128,58 g/mol
Kokpunkt: 176…178 °C
Smältpunkt: –70 °C
Densitet: 1,350
Flampunkt: 83 °C (181 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00007460
CAS nummer: 594-44-5
EINECS: 209-842-5
UN: 2927
ADR: 6.1,II