Ethanesulphonyl chloride 98+%
Leverantör: Thermo Scientific
Fara
215820050 215820500 215821000 215825000 215820025
ACRO215820050EA 430 SEK
ACRO215820050 ACRO215820500 ACRO215821000 ACRO215825000 ACRO215820025
Ethanesulphonyl chloride 98+%
Ethanesulphonyl chloride
Formel: C₂H₅ClO₂S
MW: 128,58 g/mol
Kokpunkt: 177 °C (760 mmHg)
Smältpunkt: –68 °C
Densitet: 1,35 g/cm³
Flampunkt: 83 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007460
CAS nummer: 594-44-5
EINECS: 209-842-5
UN: 2927
ADR: 6.1,II
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow to light brown Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.4520 to 1.4540 (20°C, 589 nm)
GC ≥98.0 %