1-Hexadecanesulphonyl chloride ≥98%
Leverantör: Thermo Fisher Scientific
Fara
A14396.09 A14396.18
A14396.09EA 1475 SEK
A14396.09 A14396.18
1-Hexadecanesulphonyl chloride ≥98%
1-Hexadecanesulphonyl chloride
Purity: 98% CAS No.: 38775-38-1 Molecular Formula: CH3(CH2)15SO2Cl Formula Weight: 324.96 Melting Point: 56-58° Beilstein Reference: 1789245 UN No.: 3261 MDL No.: MFCD00007459
Formel: C₁₆H₃₃ClO₂S
Smältpunkt: 56…58 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007459
CAS nummer: 38775-38-1
EINECS: 254-123-1
UN: 3261
ADR: 8,II