Dodekan-1-tiol ≥98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Mercaptan C12, 1-Mercaptododecane, n-Dodecanethiol, Lauryl mercaptan, n-Dodecyl mercaptan, NDM
A12741.22 A12741.36
A12741.22EA 565 SEK
A12741.22 A12741.36
Dodekan-1-tiol ≥98%
Dodekan-1-tiol
Purity: 98% CAS No.: 112-55-0 Molecular Formula: C12H26S Formula Weight: 202.40 Flash Point: 133°(271°F) Refractive Index: 1.4601 Odour: Unpleasant Melting Point: -7° Boiling Point: 276-278° Beilstein Reference: 969337 MDL No.: MFCD00004885
Formel: C₁₂H₂₆S
Kokpunkt: 276…278 °C
Smältpunkt: –7 °C
Densitet: 0,845
Flampunkt: 133 °C (271 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00004885
CAS nummer: 112-55-0
EINECS: 203-984-1
UN: 3334
ADR: 9,