Clelandreagens

Leverantör: RESEARCH ORGANICS
Varning

Synonyms: cis-2,3-Dihydroxy-1,4-butanedithiol, (R*,R*)-1,4-Dimercapto-2,3-butandiol, DTT, DL-1,4-Dithiothreitol

M-Clarity™ quality level = MQ300

RES2190D-A103X RES2190D-A105X RES2190D-A104X RES2190D-A102X
RESERES2190D-A103XEA 0 SEK
RESERES2190D-A103X RESERES2190D-A105X RESERES2190D-A104X RESERES2190D-A102X
Clelandreagens
Clelandreagens
Formel: C₄H₁₀O₂S₂
MW: 154,25 g/mol
Kokpunkt: ∼125 °C (1013 hPa)
Smältpunkt: 42…44 °C
Flampunkt: >110 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00004877
CAS nummer: 3483-12-3
EINECS: 222-468-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5022472c3f3baef75a703791c548fc6504df810f&vid=0d5f0a6f1cf199e6f97f1d391408acda68b4dbbf&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR