Caproic anhydride 97%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Hexanoic anhydride
186750500 186752500
ACRO186750500EA 445 SEK
ACRO186750500 ACRO186752500
Caproic anhydride 97%
Caproic anhydride
Formel: C₁₂H₂₂O₃
Kokpunkt: 246 °C (1013 hPa)
Smältpunkt: –40 °C
Densitet: 0,926 g/cm³
Flampunkt: 130 °C
Storage Temperature: Omgivande
MDL Number: MFCD00009509
CAS nummer: 2051-49-2
EINECS: 218-121-4
UN: 3265
ADR: 8,II
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4270 to 1.4300 (20°C, 589 nm)
Titration by coupling ≥96.0 %
HPLC ≥97.5 %
Sodium (Na) passes test