Glutaric anhydride ≥98%
Leverantör: Thermo Fisher Scientific
|
Fara
|
Synonyms:
Dihydro-2H-pyran-2,6(3H)-dione, Dihydro-2H-pyran-2,6(3H)-dione
A11152.0B
A11152.18
A11152.22
A11152.30
A11152.0BEA
7675
SEK
A11152.0B
A11152.18
A11152.22
A11152.30
Glutaric anhydride ≥98%
Glutaric anhydride
Purity: 98%
CAS No.: 108-55-4
Molecular Formula: C5H6O3
Formula Weight: 114.10
Flash Point: 178°(352°F)
Physical Form: Crystals
Density: 1.411
Sensitivity: Moisture Sensitive
Appearance: White to pale yellow
Melting Point: 52-57°
Boiling Point: 291°
Beilstein Reference: 110051
MDL No.: MFCD00006679
Formel:
C₅H₆O₃ Kokpunkt: 291 °C Smältpunkt: 52…57 °C Densitet: 1,411 Flampunkt: 178 °C (352 °F) Storage Temperature: Omgivande |
MDL Number:
MFCD00006679 CAS nummer: 108-55-4 EINECS: 203-593-6 |