1,2-Dimetoxietan ≥99% stabilised
Leverantör: Thermo Fisher Scientific
Fara
Stabilization: Stabilised med BHT/Ionol
A12986.AE A12986.0F A12986.AP
A12986.AEEA 610 SEK
A12986.AE A12986.0F A12986.AP
1,2-Dimetoxietan ≥99% stabilised
1,2-Dimetoxietan
Stabilized with BHT
Purity: 99+% CAS No.: 110-71-4 Molecular Formula: C4H10O2 Formula Weight: 90.12 Flash Point: -6°(21°F) Refractive Index: 1.3800 Odour: Ether-like Melting Point: -69° Boiling Point: 84-86° Beilstein Reference: 1209237 Merck Reference: 14,3224 UN No.: 2252 MDL No.: MFCD00008502
Formel: C₄H₁₀O₂
Kokpunkt: 84…86 °C
Smältpunkt: –69 °C
Densitet: 0,867
Flampunkt: –6 °C (21 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00008502
CAS nummer: 110-71-4
EINECS: 203-794-9
UN: 2252
ADR: 3,II
Merck Index: 14,3224