Trimethyl orthopropionate 97%
Leverantör: Thermo Scientific
Fara
384620250 384621000
ACRO384620250EA 1750 SEK
ACRO384620250 ACRO384621000
Trimethyl orthopropionate 97%
Trimethyl orthopropionate
Formel: C₆H₁₄O₃
MW: 134,18 g/mol
Kokpunkt: 121…122 °C (760 mmHg)
Densitet: 0,944 g/cm³
Flampunkt: 19 °C
Storage Temperature: Omgivande
MDL Number: MFCD00043305
CAS nummer: 24823-81-2
UN: 3271
ADR: 3,II
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3970 to 1.3990 (20°C, 589 nm)
GC ≥96.0 %