Dietylamin ≥99%
Leverantör: Thermo Fisher Scientific
Fara
A11716.0F A11716.AE A11716.AP
A11716.0FEA 1650 SEK
A11716.0F A11716.AE A11716.AP
Dietylamin ≥99%
Dietylamin
Purity: 99+% CAS No.: 109-89-7 Molecular Formula: C4H11N Formula Weight: 73.14 Flash Point: -28°(-18°F) Refractive Index: 1.3850 Odour: Ammonia-like Melting Point: -50° Boiling Point: 54-55° Beilstein Reference: 605268 Merck Reference: 14,3111 UN No.: 1154 MDL No.: MFCD00009032
Formel: (C₂H₅)₂NH
Kokpunkt: 54…55 °C
Smältpunkt: –50 °C
Densitet: 0,706
Flampunkt: –28 °C (–18 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00009032
CAS nummer: 109-89-7
EINECS: 203-716-3
UN: 1154
ADR: 3,II
Merck Index: 14,3111
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3c054fa528239af221505ef08b7c0d2503c9d1d7&vid=e1e59ef071f0c580f9eb9716f4801d314ff824c3&prid=web&clid=SAPDAM