1,3-Diamino-2-propanol 97%
Leverantör: Thermo Scientific
Varning
Synonyms: 1,3-Diamino-2-hydroxypropane, 1,3-Diaminopropan-2-ol
A19618.18 A19618.09 A19618.30
A19618.18EA 3825 SEK
A19618.18 A19618.09 A19618.30
1,3-Diamino-2-propanol 97%
1,3-Diamino-2-propanol
Purity: 97% CAS No.: 616-29-5 Molecular Formula: C3H10N2O Formula Weight: 90.13 Flash Point: 110°(230°F) Melting Point: 40-45° Boiling Point: 235° Beilstein Reference: 741859 MDL No.: MFCD00008142
Formel: C₃H₁₀N₂O
MW: 90,13 g/mol
Kokpunkt: 235 °C
Smältpunkt: 40…45 °C
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00008142
CAS nummer: 616-29-5
EINECS: 210-474-2