1,3-Diamino-2-propanol 95%
Leverantör: Thermo Scientific
Varning
Synonyms: 1,3-Diamino-2-hydroxypropane, 1,3-Diaminopropan-2-ol
112220050 112220250
ACRO112220050EA 1050 SEK
ACRO112220050 ACRO112220250
1,3-Diamino-2-propanol 95%
1,3-Diamino-2-propanol
Formel: C₃H₁₀N₂O
MW: 90,13 g/mol
Kokpunkt: 114 °C (5 mmHg)
Smältpunkt: 39…45 °C
Flampunkt: >110 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00008142
CAS nummer: 616-29-5
EINECS: 210-474-2
Order Now
Specifikation testresultat
Appearance White to yellow Low melting solid
Infrared spectrum Conforms
Titration with HClO4 ≥94 %
HPLC ≥94 %