1,10-Diaminodecane 97%
Leverantör: Thermo Scientific
Fara
Synonyms: Decane-1,10-diamine
112130250 112130050
ACRO112130250EA 3175 SEK
ACRO112130250 ACRO112130050
1,10-Diaminodecane 97%
1,10-Diaminodecane
Formel: C₁₀H₂₄N₂
MW: 172,31 g/mol
Kokpunkt: 140 °C (12 mmHg)
Smältpunkt: 62…63 °C
Storage Temperature: Omgivande
MDL Number: MFCD00008151
CAS nummer: 646-25-3
EINECS: 211-471-9
UN: 3259
ADR: 8,II
Order Now
Specifikation testresultat
Appearance White to yellow Crystalline powder or flakes and/or chunks
Infrared spectrum Conforms
Melting point 59 to 64 °C
GC ≥96.0 %