1-Octyl isothiocyanate

Leverantör: Thermo Scientific

Synonyms: Isothiocyanic acid n-Octyl ester, n-Octyl Isothiocyanate

TL00527EA
MAYBTL00527EAEA 0 SEK
MAYBTL00527EA
1-Octyl isothiocyanate
1-Octyl isothiocyanate
Formel: C₉H₁₇NS
MW: 171,31 g/mol
Kokpunkt: 123 °C (12 mmHg)
Densitet: 0,88 g/cm³ (20 °C)
MDL Number: MFCD00041139
CAS nummer: 4430-45-9
EINECS: 224-625-5
UN: 2922
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR