1-Octyl isothiocyanate ≥97%
Leverantör: Thermo Scientific
Fara
Synonyms: Isothiocyanic acid n-Octyl ester, n-Octyl Isothiocyanate
A14501.06 A14501.14
A14501.06EA 1050 SEK
A14501.06 A14501.14
1-Octyl isothiocyanate ≥97%
1-Octyl isothiocyanate
Purity: 97% CAS No.: 4430-45-9 Molecular Formula: CH3(CH2)7NCS Formula Weight: 171.31 Flash Point: 69°(156°F) Refractive Index: 1.4900 Odour: Pungent Boiling Point: 124-126°/7mm Beilstein Reference: 1760783 UN No.: 2922 MDL No.: MFCD00041139
Formel: C₉H₁₇NS
MW: 171,31 g/mol
Kokpunkt: 124…126 °C (7 mmHg)
Densitet: 0,880
Flampunkt: 69 °C (156 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00041139
CAS nummer: 4430-45-9
EINECS: 224-625-5
UN: 2922
ADR: 8,III